Anti-Oxytocin Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB67457-100EA 11425 NOK
ABCAAB67457-100
Anti-Oxytocin Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Oxytocin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human