Anti-ZBTB37 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67508-50EA 8050 NOK
ABCAAB67508-50
Anti-ZBTB37 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ZBTB37 (zinc finger and BTB domain containing 37)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human