Anti-ACTL7A Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67546-50EA 9475 NOK
ABCAAB67546-50
Anti-ACTL7A Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ACTL7A (actin-like 7A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human