Anti-SMARCAD1 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67548-50EA 11400 NOK
ABCAAB67548-50
Anti-SMARCAD1 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SMARCAD1 (SWI/SNF-related, matrix-associated actin-dependent regulator of chromatin, subfamily a, containing DEAD/H box 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse