Anti-EPB41L4A Mouse polyclonal antibody unconjugated
Leverandør: Abcam
ABCAAB67551-50EA 8050 NOK
ABCAAB67551-50
Anti-EPB41L4A Mouse polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: EPB41L4A (erythrocyte membrane protein band 4.1 like 4A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human