Anti-EDIL3 Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB67573-50EA 8500 NOK
ABCAAB67573-50
Anti-EDIL3 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: EDIL3 (EGF-like repeats and discoidin I-like domains 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR