Anti-PRRX1 Mouse polyclonal antibody unconjugated

ABCAAB67631-50EA 7600 NOK
ABCAAB67631-50
Anti-PRRX1 Mouse polyclonal antibody unconjugated
Type: Primary
Antigen: PRRX1 (paired related homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR