Anti-ABLIM3 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67721-50EA 10900 NOK
ABCAAB67721-50
Anti-ABLIM3 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ABLIM3 (actin binding LIM protein family, member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse