Anti-HESX1 Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB67728-50EA 11400 NOK
ABCAAB67728-50
Anti-HESX1 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HESX1 (HESX homeobox 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR