Anti-CDC42EP4 Mouse polyclonal antibody unconjugated

ABCAAB67780-50EA 7175 NOK
ABCAAB67780-50
Anti-CDC42EP4 Mouse polyclonal antibody unconjugated
Type: Primary
Antigen: CDC42EP4 (CDC42 effector protein (Rho GTPase binding) 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR