Anti-MED20 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67800-50EA 10900 NOK
ABCAAB67800-50
Anti-MED20 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MED20 (mediator complex subunit 20)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human