Anti-DUSP27 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67829-50EA 10900 NOK
ABCAAB67829-50
Anti-DUSP27 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: DUSP27 (dual specificity phosphatase 27 (putative))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human