Anti-ACVR1C Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67921-50EA 11475 NOK
ABCAAB67921-50
Anti-ACVR1C Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ACVR1C (activin A receptor, type IC)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human