Anti-CLK4 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67936-50EA 11475 NOK
ABCAAB67936-50
Anti-CLK4 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: CLK4 (CDC-like kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human