Anti-SLC26A6 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB67959-50EA 11450 NOK
ABCAAB67959-50
Anti-SLC26A6 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: SLC26A6 (solute carrier family 26 (anion exchanger), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human