Anti-Kindlin Rabbit Polyclonal Antibody
Leverandør: Abcam
ABCAAB68041-100EA 12100 NOK
ABCAAB68041-100
Anti-Kindlin Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Kindlin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse