Anti-FBXL5 Mouse polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB68069-50EA 8425 NOK
ABCAAB68069-50
Anti-FBXL5 Mouse polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: FBXL5 (F-box and leucine-rich repeat protein 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR