Anti-BCKDHA Mouse polyclonal antibody unconjugated

ABCAAB68094-50EA 7600 NOK
ABCAAB68094-50
Anti-BCKDHA Mouse polyclonal antibody unconjugated
Type: Primary
Antigen: BCKDHA (branched chain keto acid dehydrogenase E1, alpha polypeptide)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR