Anti-OLIG1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB68105-100EA 10275 NOK
ABCAAB68105-100
Anti-OLIG1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: OLIG1 (oligodendrocyte transcription factor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR