Anti-L3MBTL3 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB68117-50EA 10900 NOK
ABCAAB68117-50
Anti-L3MBTL3 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: L3MBTL3 (l(3)mbt-like 3 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human