Anti-STAT4 Rabbit Monoclonal Antibody [clone: EP1900Y]

Leverandør: Abcam
ABCAAB68156-100EA 10450 NOK
ABCAAB68156-100
Anti-STAT4 Rabbit Monoclonal Antibody [clone: EP1900Y]
Antistoffer
Type: Primary
Antigen: STAT4 (signal transducer and activator of transcription 4)
Clonality: monoclonal
Clone: EP1900Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR