Anti-TLR1 Rabbit Monoclonal Antibody [clone: EPR2075]
Leverandør: Abcam
ABCAAB68158-100EA 11575 NOK
ABCAAB68158-100
Anti-TLR1 Rabbit Monoclonal Antibody [clone: EPR2075]
Antistoffer
Type: Primary
Antigen: TLR1 (toll-like receptor 1)
Clonality: monoclonal
Clone: EPR2075
Conjugation: unconjugated
Epitope: extracellular domain
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå