Anti-eIF4EBP1 + eIF4EBP2 + eIF4EBP3 Rabbit monoclonal antibody unconjugated [clone: EPR2169Y]

Leverandør: Abcam
ABCAAB68187-100EA 11725 NOK
ABCAAB68187-100
Anti-eIF4EBP1 + eIF4EBP2 + eIF4EBP3 Rabbit monoclonal antibody unconjugated [clone: EPR2169Y]
Antistoffer
Type: Primary
Antigen: eIF4EBP1 + eIF4EBP2 + eIF4EBP3
Clonality: monoclonal
Clone: EPR2169Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR