Anti-ACTN3 Rabbit Monoclonal Antibody [clone: EP2531Y]

Leverandør: Abcam
ABCAAB68204-100EA 10600 NOK
ABCAAB68204-100
Anti-ACTN3 Rabbit Monoclonal Antibody [clone: EP2531Y]
Antistoffer
Type: Primary
Antigen: ACTN3 (actinin, alpha 3 (gene/pseudogene))
Clonality: monoclonal
Clone: EP2531Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR