Anti-PTTG1IP Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB68208-50EA 11400 NOK
ABCAAB68208-50
Anti-PTTG1IP Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PTTG1IP (pituitary tumor-transforming 1 interacting protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR