Anti-BCR Rabbit monoclonal antibody unconjugated [clone: EPR576Y]
Leverandør: Abcam
ABCAAB68216-100EA 10625 NOK
ABCAAB68216-100
Anti-BCR Rabbit monoclonal antibody unconjugated [clone: EPR576Y]
Antistoffer
Type: Primary
Antigen: BCR (breakpoint cluster region)
Clonality: monoclonal
Clone: EPR576Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå