Anti-BTK Rabbit Monoclonal Antibody [clone: EP420Y]

Leverandør: Abcam
ABCAAB68217-100EA 12600 NOK
ABCAAB68217-100
Anti-BTK Rabbit Monoclonal Antibody [clone: EP420Y]
Antistoffer
Type: Primary
Antigen: BTK (Bruton agammaglobulinemia tyrosine kinase)
Clonality: monoclonal
Clone: EP420Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR