Anti-BBS2 Mouse polyclonal antibody unconjugated

Leverandør: Abcam
ABCAAB68223-50EA 8500 NOK
ABCAAB68223-50
Anti-BBS2 Mouse polyclonal antibody unconjugated
Antistoffer
Type: Primary
Antigen: BBS2 (Bardet-Biedl syndrome 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR