Anti-UBE3C Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB68225-50EA 8450 NOK
ABCAAB68225-50
Anti-UBE3C Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: UBE3C (ubiquitin protein ligase E3C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR