Anti-CHEK2 Rabbit Monoclonal Antibody [clone: EPR916Y]
Leverandør: Abcam
ABCAAB68267-100EA 10625 NOK
ABCAAB68267-100
Anti-CHEK2 Rabbit Monoclonal Antibody [clone: EPR916Y]
Antistoffer
Type: Primary
Antigen: CHEK2 (checkpoint kinase 2)
Clonality: monoclonal
Clone: EPR916Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå