Anti-HSPD1 Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB68416-100EA 8625 NOK
ABCAAB68416-100
Anti-HSPD1 Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: HSPD1 (heat shock 60kDa protein 1 (chaperonin))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR