Anti-FCGR2A Rabbit monoclonal antibody unconjugated [clone: EP926Y]

Leverandør: Abcam
ABCAAB68423-100EA 11775 NOK
ABCAAB68423-100
Anti-FCGR2A Rabbit monoclonal antibody unconjugated [clone: EP926Y]
Antistoffer
Type: Primary
Antigen: FCGR2A (Fc fragment of IgG, low affinity IIa, receptor (CD32))
Clonality: monoclonal
Clone: EP926Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR