Anti-GABRB2 Rabbit monoclonal antibody unconjugated [clone: EP2317Y]

Leverandør: Abcam
ABCAAB68425-100EA 10625 NOK
ABCAAB68425-100
Anti-GABRB2 Rabbit monoclonal antibody unconjugated [clone: EP2317Y]
Antistoffer
Type: Primary
Antigen: GABRB2 (gamma-aminobutyric acid (GABA) A receptor, beta 2)
Clonality: monoclonal
Clone: EP2317Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR