Anti-KRT7 Rabbit Monoclonal Antibody [clone: EP1620Y]

Leverandør: Abcam
ABCAAB68460-100EA 11350 NOK
ABCAAB68460-100
Anti-KRT7 Rabbit Monoclonal Antibody [clone: EP1620Y]
Antistoffer
Type: Primary
Antigen: KRT7 (keratin 7)
Clonality: monoclonal
Clone: EP1620Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR