Anti-FADD Rabbit Monoclonal Antibody [clone: EPR1816Y]
Leverandør: Abcam
ABCAAB68475-100EA 10625 NOK
ABCAAB68475-100
Anti-FADD Rabbit Monoclonal Antibody [clone: EPR1816Y]
Antistoffer
Type: Primary
Antigen: FADD (Fas (TNFRSF6)-associated via death domain)
Clonality: monoclonal
Clone: EPR1816Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå