Anti-ERBB4 Rabbit Monoclonal Antibody [clone: EPR2271Y]
Leverandør: Abcam
ABCAAB68479-100EA 10625 NOK
ABCAAB68479-100
Anti-ERBB4 Rabbit Monoclonal Antibody [clone: EPR2271Y]
Antistoffer
Type: Primary
Antigen: ERBB4 (v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 4)
Clonality: monoclonal
Clone: EPR2271Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå