Anti-IRF3 Rabbit Monoclonal Antibody [clone: EPR2418Y]

Leverandør: Abcam
ABCAAB68481-100EA 12825 NOK
ABCAAB68481-100
Anti-IRF3 Rabbit Monoclonal Antibody [clone: EPR2418Y]
Antistoffer
Type: Primary
Antigen: IRF3 (interferon regulatory factor 3)
Clonality: monoclonal
Clone: EPR2418Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR