Anti-Clostridia spp Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

ABCAAB68500-500EA 7175 NOK
ABCAAB68500-500
Anti-Clostridia spp Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Type: Primary
Antigen: Clostridia spp
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR