Anti-Chlamydia trachomatis MOMP Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Abcam
ABCAAB68509-500EA 8450 NOK
ABCAAB68509-500
Anti-Chlamydia trachomatis MOMP Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Primary
Antigen: Chlamydia trachomatis MOMP
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR