Anti-FAN1 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB68572-50EA 11475 NOK
ABCAAB68572-50
Anti-FAN1 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: FAN1 (FANCD2/FANCI-associated nuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human