Anti-Hepatitis A Virus Goat Polyclonal Antibody
Leverandør: Abcam
ABCAAB68579-500EA 11400 NOK
ABCAAB68579-500
Anti-Hepatitis A Virus Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Hepatitis A Virus
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestill nå