Anti-Listeria monocytogenes Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Abcam
ABCAAB68592-500EA 11400 NOK
ABCAAB68592-500
Anti-Listeria monocytogenes Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: Listeria monocytogenes
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR