Anti-HIV1 gp41 Goat Polyclonal Antibody (Biotin)

Leverandør: Abcam
ABCAAB68623-500EA 11400 NOK
ABCAAB68623-500
Anti-HIV1 gp41 Goat Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: HIV1 gp41
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR