Anti-SMC3 Rabbit polyclonal antibody unconjugated

ABCAAB68655-100EA 7425 NOK
ABCAAB68655-100
Anti-SMC3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: SMC3 (structural maintenance of chromosomes 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR