Anti-AVP Rabbit Polyclonal Antibody

Leverandør: Abcam
ABCAAB68669-100EA 11900 NOK
ABCAAB68669-100
Anti-AVP Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: AVP (arginine vasopressin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR