Anti-TSEN34 Mouse Polyclonal Antibody

Leverandør: Abcam
ABCAAB68868-50EA 10900 NOK
ABCAAB68868-50
Anti-TSEN34 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TSEN34 (TSEN34 tRNA splicing endonuclease subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR