Anti-ACSS3 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB68927-50EA 10900 NOK
ABCAAB68927-50
Anti-ACSS3 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: ACSS3 (acyl-CoA synthetase short-chain family member 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human