Anti-Staphylococcus aureus Rabbit Polyclonal Antibody (FITC (Fluorescein))
Leverandør: Abcam
ABCAAB68950-500EA 11400 NOK
ABCAAB68950-500
Anti-Staphylococcus aureus Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Type: Primary
Antigen: Staphylococcus aureus
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå