Anti-WSB1 Mouse polyclonal antibody unconjugated

ABCAAB68953-50EA 7175 NOK
ABCAAB68953-50
Anti-WSB1 Mouse polyclonal antibody unconjugated
Type: Primary
Antigen: WSB1 (WD repeat and SOCS box containing 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR