Anti-TINAGL1 Mouse Polyclonal Antibody
Leverandør: Abcam
ABCAAB69036-50EA 10900 NOK
ABCAAB69036-50
Anti-TINAGL1 Mouse Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: TINAGL1 (tubulointerstitial nephritis antigen-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human